1-(3-Bromobenzyl)piperazine
Supplier: Matrix Scientific
027771-500MG
101824-824EA
193.85
USD
101824-824
1-(3-Bromobenzyl)piperazine
1-(3-Bromobenzyl)piperazine
Formula:
C₁₁H₁₅BrN₂ |
MDL Number:
MFCD02211309 CAS Number: 423154-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...