3-Phenylbenzoic acid

Synonyms: 1,1'-Biphenyl-3-carboxylic acidBiphenyl-3-carboxylic acid3-Biphenylcarboxylic acid[1,1'-Biphenyl]-3-carboxylic acid

027211-500MG
101823-738EA 136.43 USD
101823-738
3-Phenylbenzoic acid
3-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD00045846
CAS Number: 716-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR