2-Phenylpiperidine

Supplier: Matrix Scientific

027113-500MG
101823-522EA 78.46 USD
101823-522
2-Phenylpiperidine
2-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
MDL Number: MFCD00270125
CAS Number: 3466-80-6
EINECS: 222-422-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR