2-Amino-5-chloro-3-fluoropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-3-fluoropyridin-2-amine5-Chloro-3-fluoro-2-pyridylamine

026727-5G
101822-734EA 73.85 USD
101822-734
2-Amino-5-chloro-3-fluoropyridine ≥98%
2-Amino-5-chloro-3-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₄ClFN₂
MW: 146.55 g/mol
Melting Pt: 95…97 °C
MDL Number: MFCD09878434
CAS Number: 246847-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR