2,4,6-Trichloropyrimidine ≥98%
Supplier: Matrix Scientific
|
Warning
|
026725-25G
026725-100G
101822-728EA
36.92
USD
101822-728
101822-730
2,4,6-Trichloropyrimidine ≥98%
2,4,6-Trichloropyrimidine
Hazard code(s): Corrosive, Lachrymator
Formula:
C₄HCl₃N₂ MW: 183.42 g/mol Boiling Pt: 213…215 °C Melting Pt: 23…25 °C Density: 1.595 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00006063 CAS Number: 3764-01-0 EINECS: 223-183-0 |
Specification Test Results
Boiling point | 213-215°C |
Melting point | 23-25°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...