5-Bromo-2-hydroxypyridine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Bromo-6-pyridinol5-Bromo-2-pyridinol5-Bromopyridin-2-ol3-Bromo-6-hydroxypyridine

026298-100G
101822-028EA 181.54 USD
101822-028
5-Bromo-2-hydroxypyridine ≥98%
5-Bromo-2-hydroxypyridine

Hazard code(s): Irritant

Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR