2-Chloro-5-fluoro-3-iodopyridine

Supplier: Matrix Scientific

026177-250MG
101821-760EA 427.69 USD
101821-760
2-Chloro-5-fluoro-3-iodopyridine
2-Chloro-5-fluoro-3-iodopyridine
Formula: C₅H₂ClFIN
MW: 257.43 g/mol
MDL Number: MFCD03095255
CAS Number: 884494-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR