2-Isopropoxy-3-pyridineboronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 2-Isopropoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine2-Isopropoxypyridine-3-boronic acid pinacol ester2-Isopropoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

026132-500MG
101821-668EA 74.75 USD
101821-668
2-Isopropoxy-3-pyridineboronic acid pinacol ester
2-Isopropoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
MDL Number: MFCD07366722
CAS Number: 848243-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR