2-Ethoxy-3-pyridineboronic acid pinacol ester

Supplier: Matrix Scientific

Synonyms: 2-Ethoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine2-Ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Ethoxypyridine-3-boronic acid pinacol ester

026131-500MG
101821-666EA 69.23 USD
101821-666
2-Ethoxy-3-pyridineboronic acid pinacol ester
2-Ethoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249.12 g/mol
MDL Number: MFCD07366721
CAS Number: 848243-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR