1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene ≥95%

Supplier: Matrix Scientific

024956-5G 024956-25G
101819-284EA 52.31 USD
101819-284 101819-286
1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene ≥95%
1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene

Hazard code(s): Irritant

Formula: C₁₅H₂₂
MW: 202.34 g/mol
Melting Pt: 31 °C
MDL Number: MFCD00833394
CAS Number: 6683-48-3
EINECS: 229-724-7

Order Now

Specification Test Results

Melting point 31 oC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR