D-(+)-Lactamide ≥97%, ee 98%

Supplier: Matrix Scientific

Synonyms: (R)-2-Hydroxypropanamide

024802-5G
101818-930EA 86.25 USD
101818-930
D-(+)-Lactamide ≥97%, ee 98%
D(+)-Lactamide
Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00210310
CAS Number: 598-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR