2,3-Dimethylpiperazine ≥95%

Supplier: Matrix Scientific

024769-500MG
101818-864EA 209.23 USD
101818-864
2,3-Dimethylpiperazine ≥95%
2,3-Dimethylpiperazine

Hazard code(s): Irritant

Formula: C₆H₁₄N₂
Melting Pt: 82…88 °C
MDL Number: MFCD07373397
CAS Number: 84468-52-0

Order Now

Specification Test Results

Melting point 82-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR