3-Chloro-5-formylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Chloronicotinaldehyde5-Chloro-pyridine-3-carbaldehyde

024719-250MG
101818-722EA 61.54 USD
101818-722
3-Chloro-5-formylpyridine ≥98%
3-Chloro-5-formylpyridine

Hazard code(s): Irritant

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Melting Pt: 70…71 °C
MDL Number: MFCD08457773
CAS Number: 113118-82-4

Order Now

Specification Test Results

Melting point 70-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR