3-Methyl-5-phenyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

024698-1G
101818-664EA 123.08 USD
101818-664
3-Methyl-5-phenyl-1H-pyrazole ≥97%
3-Methyl-5-phenyl-1H-pyrazole

Hazard code(s): Irritant

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Melting Pt: 126…127 °C
MDL Number: MFCD00524752
CAS Number: 3347-62-4
EINECS: 222-099-1

Order Now

Specification Test Results

Melting point 126-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR