2,2-Difluoro-4-pentenoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluoropent-4-enoic acid

024692-1G
101818-654EA 150.77 USD
101818-654
2,2-Difluoro-4-pentenoic acid ≥97%
2,2-Difluoro-4-pentenoic acid

Hazard code(s): Irritant

Formula: C₅H₆F₂O₂
MW: 136.1 g/mol
Boiling Pt: 56…58 °C (3mm)
MDL Number: MFCD09800642
CAS Number: 55039-89-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 56-58°C/3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR