2-Chloro-5-methyl-3-nitropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-nitro-5-picoline

024636-25G
101818-514EA 76.92 USD
101818-514
2-Chloro-5-methyl-3-nitropyridine ≥98%
2-Chloro-5-methyl-3-nitropyridine

Hazard code(s): Irritant

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD02070020
CAS Number: 23056-40-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR