3-Amino-2-fluoro-6-methylpyridine ≥98%
Supplier: Matrix Scientific
024605-5G
024605-1G
101818-412EA
36.92
USD
101818-412
101818-414
3-Amino-2-fluoro-6-methylpyridine ≥98%
3-Amino-2-fluoro-6-methylpyridine
Hazard code(s): Irritant
Formula:
C₆H₇FN₂ |
MDL Number:
MFCD03095221 CAS Number: 374633-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...