4-Chlorobiphenyl (PCB No. 3) ≥95%

Supplier: Matrix Scientific

024600-1G
101818-398EA 60 USD
101818-398
4-Chlorobiphenyl (PCB No. 3) ≥95%
4-Chlorobiphenyl (PCB No. 3)

Hazard code(s): Irritant

Formula: C₁₂H₉Cl
MW: 188.66 g/mol
Boiling Pt: 130 °C (1mm)
Melting Pt: 77…78 °C
MDL Number: MFCD00013640
CAS Number: 2051-62-9
EINECS: 218-127-7

Order Now

Specification Test Results

Boiling point 130°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR