3-Bromo-5-methylpyridine ≥98%
Supplier: Matrix Scientific
024592-25G
024592-5G
102551-060EA
112.31
USD
102551-060
101818-392
3-Bromo-5-methylpyridine ≥98%
3-Bromo-5-methylpyridine
Hazard code(s): Irritant
Formula:
C₆H₆BrN MW: 172.02 g/mol Density: 1.49 g/cm³ |
MDL Number:
MFCD01646141 CAS Number: 3430-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...