4-Isopropoxybenzoic acid

Supplier: Matrix Scientific

024089-25G
101817-388EA 387.69 USD
101817-388
4-Isopropoxybenzoic acid
4-Isopropoxybenzoic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00044318
CAS Number: 13205-46-4
EINECS: 236-169-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR