4-Amino-N,N-dimethylbenzenesulfonamide ≥95%

Supplier: Matrix Scientific

023877-1G
101816-954EA 44.62 USD
101816-954
4-Amino-N,N-dimethylbenzenesulfonamide ≥95%
4-Amino-N,N-dimethylbenzenesulphonamide

Hazard code(s): Irritant

Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
MDL Number: MFCD00031428
CAS Number: 1709-59-7
EINECS: 216-970-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR