2-Chloro-5-fluoropyrimidine ≥98%

Supplier: Matrix Scientific

023813-25G
101816-822EA 135.38 USD
101816-822
2-Chloro-5-fluoropyrimidine ≥98%
2-Chloro-5-fluoropyrimidine

Hazard code(s): Corrosive, Irritant

Formula: C₄H₂ClFN₂
Density: 1.073 g/cm³
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR