2-Amino-4-chloropyridine ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Chloro-2-pyridylamine4-Chloropyridin-2-amine

023809-25G
101816-802EA 69.23 USD
101816-802
2-Amino-4-chloropyridine ≥97%
2-Amino-4-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 129…133 °C
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Specification Test Results

Melting point 129-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR