5-Fluoro-2-methoxypyridine ≥98%

Supplier: Matrix Scientific

023767-5G
101816-680EA 60 USD
101816-680
5-Fluoro-2-methoxypyridine ≥98%
5-Fluoro-2-methoxypyridine

Hazard code(s): Flammable, Store in Freezer

Formula: C₆H₆FNO
Boiling Pt: 137…140 °C
MDL Number: MFCD03095252
CAS Number: 51173-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR