2-Chloro-4-(trifluoromethyl)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

023744-500MG
101816-620EA 49.23 USD
101816-620
2-Chloro-4-(trifluoromethyl)benzenesulfonamide ≥97%
2-Chloro-4-(trifluoromethyl)benzenesulphonamide

Hazard code(s): Irritant

Formula: C₇H₅ClF₃NO₂S
MW: 259.64 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00179384
CAS Number: 146533-47-3

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR