Methyl-6-methoxypicolinate ≥98%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-methoxypyridine-2-carboxylate
, 6-Methoxy-pyridine-2-carboxylic acid methyl ester
, Methyl 6-methoxypicolinate
023731-250MG
101816-588EA
273.85
USD
101816-588
Methyl-6-methoxypicolinate ≥98%
Methyl 6-methoxypicolinate
Hazard code(s): Irritant
Formula:
C₈H₉NO₃ MW: 167.1600 g/mol Melting Pt: 38.5…39.5 °C |
MDL Number:
MFCD07779517 CAS Number: 26256-72-4 |
Specification Test Results
Melting point | 38.5-39.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...