2-Amino-6-chloropyridine ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 6-Chloropyridin-2-amine6-Chloro-2-pyridylamine

023713-25G
101816-550EA 44.62 USD
101816-550
2-Amino-6-chloropyridine ≥97%
2-Amino-6-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00234068
CAS Number: 45644-21-1

Order Now

Specification Test Results

Melting point 70-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR