1-(2-Methoxyethyl)piperazine

Supplier: Matrix Scientific

021600-25G 021600-5G
101812-398EA 193.85 USD
101812-398 101812-396
1-(2-Methoxyethyl)piperazine
1-(2-Methoxyethyl)piperazine
Formula: C₇H₁₆N₂O
MW: 144.22 g/mol
MDL Number: MFCD00191214
CAS Number: 13484-40-7
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR