(S)-(+)-2-Amino-1-propanol

Supplier: Matrix Scientific
Danger

Synonyms: L(+)-AlaninolL-AlaninolL(+)-2-Amino-1-propanol(S)-2-Aminopropan-1-ol

021579-25G
101812-354EA 30.77 USD
101812-354
(S)-(+)-2-Amino-1-propanol
(S)-(+)-2-Amino-1-propanol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 173 °C (1013 hPa)
Density: 0.955 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064412
CAS Number: 2749-11-3
EINECS: 220-388-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR