2,3-Dibromo-5-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3-Dibromo-5-methylpyridine

021416-5G
101812-040EA 23.08 USD
101812-040
2,3-Dibromo-5-picoline ≥98%
2,3-Dibromo-5-picoline

Hazard code(s): Irritant

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD04112574
CAS Number: 29232-39-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR