1,3,5,7-Cyclooctatetraene ≥98%

Supplier: Matrix Scientific

Synonyms: CyclooctatetraeneCOT

021368-5G
101811-868EA 292.31 USD
101811-868
1,3,5,7-Cyclooctatetraene ≥98%
1,3,5,7-Cyclooctatetraene

Inhibited with 0.1% hydroquinone


Hazard code(s): Flammable, Refrigerate

Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 142…143 °C
Melting Pt: –5 °C
Density: 0.925 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00004161
CAS Number: 629-20-9
EINECS: 211-080-3
UN: 2358
ADR: 3,II

Order Now

Specification Test Results

Boiling point 142-143°C
Density 0.925
ND20 1.537

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR