4-Methoxyphenylhydrazinium chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methoxyphenylhydrazine hydrochloride

021131-100G 021131-25G
101811-410EA 207.69 USD
101811-410 101811-408
4-Methoxyphenylhydrazinium chloride ≥97%
4-Methoxyphenylhydrazinium chloride

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₁₁ClN₂O
MW: 174.63 g/mol
Melting Pt: 161 °C (dec.)
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3

Order Now

Specification Test Results

Melting point 161°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR