4-Bromo-2,3-difluorophenol ≥97%
Supplier: Matrix Scientific
|
Warning
|
021128-100G
021128-25G
021128-5G
101811-402EA
41.54
USD
101811-402
101811-406
101811-404
4-Bromo-2,3-difluorophenol ≥97%
4-Bromo-2,3-difluorophenol
Hazard code(s): Irritant
Formula:
C₆H₃BrF₂O MW: 208.99 g/mol |
MDL Number:
MFCD08061907 CAS Number: 144292-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...