2-Bromopyrimidine ≥95%

Supplier: Matrix Scientific
Warning

020965-100G
101811-120EA 412.31 USD
101811-120
2-Bromopyrimidine ≥95%
2-Bromopyrimidine

Hazard code(s): Irritant

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 62…64 °C (1.5mm)
Melting Pt: 55…57 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

Specification Test Results

Boiling point 62-64°C/1.5
Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR