1-Benzhydrylpiperazine

Supplier: Matrix Scientific

Synonyms: 1-(Diphenylmethyl)piperazine

020907-100G 020907-25G
101810-990EA 32.31 USD
101810-990 101810-992
1-Benzhydrylpiperazine
1-Benzhydrylpiperazine

Hazard code(s): Irritant

Formula: C₁₇H₂₀N₂
Melting Pt: 85…88 °C
MDL Number: MFCD00038379
CAS Number: 841-77-0
EINECS: 212-667-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR