Dimethylmalonic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethylmalonic acid

020747-100G 020747-25G 020747-5G
101810-696EA 21.54 USD
101810-696 101810-698 101810-700
Dimethylmalonic acid ≥95%
Dimethylmalonic acid

Hazard code(s): Irritant

Formula: C₅H₈O₄
MW: 132.12 g/mol
Melting Pt: 192 °C (dec.)
MDL Number: MFCD00004193
CAS Number: 595-46-0
EINECS: 209-867-1

Order Now

Specification Test Results

Melting point 192°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR