5-Bromo-2-hydroxy-3-nitropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-3-nitro-2-pyridinol

020586-5G
101810-370EA 26.15 USD
101810-370
5-Bromo-2-hydroxy-3-nitropyridine ≥97%
5-Bromo-2-hydroxy-3-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₃BrN₂O₃
Melting Pt: 148…150 °C
MDL Number: MFCD00023473
CAS Number: 15862-34-7
EINECS: 239-989-0

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR