3-Amino-2-chloro-4-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methylpyridin-3-amine

020582-25G
101810-352EA 29.23 USD
101810-352
3-Amino-2-chloro-4-methylpyridine ≥98%
3-Amino-2-chloro-4-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD00673152
CAS Number: 133627-45-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR