5-Methyl-2-furoyl chloride

Supplier: Matrix Scientific

Synonyms: 5-Methyl-furan-2-carbonyl chloride

020334-500MG
101809-834EA 76.92 USD
101809-834
5-Methyl-2-furoyl chloride
5-Methyl-2-furoyl chloride
Formula: C₆H₅ClO₂
MDL Number: MFCD03421416
CAS Number: 14003-11-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR