1-(2-Bromoethoxy)-3,5-dimethylbenzene

Supplier: Matrix Scientific

020316-500MG
101809-798EA 280 USD
101809-798
1-(2-Bromoethoxy)-3,5-dimethylbenzene
1-(2-Bromoethoxy)-3,5-dimethylbenzene
Formula: C₁₀H₁₃BrO
MDL Number: MFCD02030859
CAS Number: 37136-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR