2-Chloro-5-fluorobenzaldehyde

Supplier: Matrix Scientific

018354-5G
101806-024EA 38.46 USD
101806-024
2-Chloro-5-fluorobenzaldehyde
2-Chloro-5-fluorobenzaldehyde
Formula: C₇H₄ClFO
MDL Number: MFCD03788511
CAS Number: 84194-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR