3-Bromo-5-fluoropyridine ≥97%

Supplier: Matrix Scientific

018350-5G
101806-008EA 18.46 USD
101806-008
3-Bromo-5-fluoropyridine ≥97%
3-Bromo-5-fluoropyridine

Hazard code(s): Toxic, Store in Freezer

Formula: C₅H₃BrFN
Melting Pt: 24…28 °C
MDL Number: MFCD04112555
CAS Number: 407-20-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR