trans-2-Fluorocyclohexanol ≥97%

Supplier: Matrix Scientific

018348-250MG
101805-998EA 69.23 USD
101805-998
trans-2-Fluorocyclohexanol ≥97%
trans-2-Fluorocyclohexanol

Hazard code(s): Irritant

Formula: C₆H₁₁FO
MW: 118.15 g/mol
Boiling Pt: 60 °C (12mm)
MDL Number: MFCD08146648
CAS Number: 656-60-0

Order Now

Specification Test Results

Boiling point 60°C/12mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR