2-Bromo-5-fluoro-6-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-fluoro-6-picoline

018333-5G
101805-946EA 110.77 USD
101805-946
2-Bromo-5-fluoro-6-methylpyridine ≥98%
2-Bromo-5-fluoro-6-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD03095064
CAS Number: 374633-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR