2-Bromo-5-hydroxypyridine ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2-Bromo-5-pyridinol6-Bromopyridin-3-ol

018332-5G
101805-940EA 35.38 USD
101805-940
2-Bromo-5-hydroxypyridine ≥98%
2-Bromo-5-hydroxypyridine

Hazard code(s): Irritant

Formula: C₅H₄BrNO
MW: 174 g/mol
MDL Number: MFCD04114184
CAS Number: 55717-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR