α,α-Difluorophenylacetic acid

Supplier: Matrix Scientific
Warning

Synonyms: α,α-Difluorobenzeneacetic acid

018321-5G
101805-914EA 69.23 USD
101805-914
α,α-Difluorophenylacetic acid
α,α-Difluorophenylacetic acid
Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
MDL Number: MFCD00498146
CAS Number: 360-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR