4-Chloroisatin

Supplier: Matrix Scientific

Synonyms: 4-Chloroindoline-2,3-dione

018300-5G
101805-868EA 23.08 USD
101805-868
4-Chloroisatin
4-Chloroisatin

Hazard code(s): Irritant

Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
Melting Pt: 258 °C
MDL Number: MFCD00129178
CAS Number: 6344-05-4

Order Now

Specification Test Results

Melting point 258 °CC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR