1,3-Difluoro-5-nitrobenzene ≥98%

Supplier: Matrix Scientific
Warning

018292-25G
101805-848EA 269.23 USD
101805-848
1,3-Difluoro-5-nitrobenzene ≥98%
1,3-Difluoro-5-nitrobenzene

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 176…177 °C
Melting Pt: 17 °C
Density: 1.407 g/cm³
MDL Number: MFCD00012142
CAS Number: 2265-94-3
EINECS: 218-867-0

Order Now

Specification Test Results

Boiling point 176-177°C
Melting point 17°C
Density 1.407
ND20 1.499

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR