6-Methoxypicolinic acid

Supplier: Matrix Scientific

Synonyms: 6-Methoxypyridine-2-carboxylic acid6-Methoxy-2-pyridinecarboxylic acid

018259-5G
101805-758EA 75.38 USD
101805-758
6-Methoxypicolinic acid
6-Methoxypicolinic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD06800962
CAS Number: 26893-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR