2-Bromonicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-pyridinecarboxylic acid2-Bromopyridine-3-carboxylic acid

018224-25G
101805-676EA 67.69 USD
101805-676
2-Bromonicotinic acid ≥97%
2-Bromonicotinic acid

Hazard code(s): Irritant

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD01646068
CAS Number: 35905-85-2

Order Now

Specification Test Results

Melting point 200°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR